missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Sphingosine Kinase 2/SPHK2 Protein

Product Code. 18168200
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18168200 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18168200 Supplier R&D Systems™ Supplier No. 6058SK010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Sphingosine Kinase 2/SPHK2 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Sphingosine Kinase 2/SPHK2 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT, Glycerol and Tween™ 20.
Gene ID (Entrez) 56632
Molecular Weight (g/mol) 67 kDa
Name Sphingosine Kinase 2/SPHK2
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Sphingosine Kinase 2/SPHK2 protein Ala2-Ala617,with an N-terminal Met and an N-terminal 7-His tag Accession &Num; Q9JIA7
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.