missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Spermine oxidase Protein

A DNA sequence encoding the Mus musculus (Mouse) Spermine oxidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6701-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095230

  • £2480.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001171304.1
Spermine oxidase Protein
Research Use Only
Smox
Recombinant Protein
Yeast
MQSCESSGDS ADDPLSRGLR RRGQPRVVVI GAGLAGLAAA RALLEQGFTD VTVLEASSHI GGRVQSVRLG DTTFELGATW IHGSHGNPIY QLAEANGLLE ETTDGERSVG RISLYSKNGV ACYLTNRGCR IPKDVVEEFS DLYNEVYNMT QEFFRHGKPV NAESQNSVGV FTREKVRNRI RDDPDDTEAT KRLKLAMIQQ YLKVESCESS SHSIDEVSLS AFGEWTEIPG AHHIIPSGFM RVVELLAEGI PPHVIQLGKP VRCIHWDQAS AHPRGPEIEP RGEGDHNHDT GEGGQSGENP QQGRWDEDEP WPVVVECEDC EVIPADHVIV TVSLGVLKRQ YTSFFRPCLP TEKVAAIHRL GIGTTDKIFL EFEEPFWGPE CNSLQFVWED EAESCTLTYP PELWYRKICG FDVLYPPERY GHVLSGWICG EEALVMERCD DEAVAEICTE MLRQFTGNPN IPKPRRILRS AWGSNPYFRG SYSYTQVGSS GADVEKLAKP LPYTESSKTA PMQVLFSGEA THRKYYSTTH GALLSGQREA ARLIEMYRDL FQQGP
Greater than 90% as determined by SDS-PAGE.
228608
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers