missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse SOST/Sclerostin Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1589-ST-025/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18370548

  • £399.56 / 25µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse SOST/Sclerostin Protein is derived from NS0. The Recombinant Mouse SOST/Sclerostin Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
22 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
74499
SOST/Sclerostin
Mouse myeloma cell line,NS0-derived mouse SOST/Sclerostin protein Gln24-Tyr211 with an N-terminal 7-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers