missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Sonic Hedgehog/Shh, N-Terminus Protein

Product Code. 18128240
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Lyophilized from a 0.2μm filtered solution in PBS, Trehalose and with BSA as a carrier protein.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18128240 Lyophilized from a 0.2μm filtered solution in PBS, Trehalose and with BSA as a carrier protein. 25µg
18399477 Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18128240 Supplier R&D Systems™ Supplier No. 461SH025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Sonic Hedgehog/Shh, N-Terminus Protein is derived from E. coli. The Recombinant Mouse Sonic Hedgehog/Shh, N-Terminus Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS, Trehalose and with BSA as a carrier protein.
Gene ID (Entrez) 20423
Molecular Weight (g/mol) 20 kDa
Name Sonic Hedgehog/Shh N-Terminus
Quantity 25 μg
Source E. coli-derived mouse Sonic Hedgehog/Shh protein Cys25-Gly198,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.