missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SOD1 / Superoxide Dismutase Protein

A DNA sequence encoding the Mus musculus (Mouse) SOD1 / Superoxide Dismutase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6715-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095320

  • £2480.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_035564.1
SOD1 / Superoxide Dismutase Protein
Research Use Only
sod1
Recombinant Protein
E. coli
AMKAVCVLKG DGPVQGTIHF EQKASGEPVV LSGQITGLTE GQHGFHVHQY GDNTQGCTSA GPHFNPHSKK HGGPADEERH VGDLGNVTAG KDGVANVSIE DRVISLSGEH SIIGRTMVVH EKQDDLGKGG NEESTKTGNA GSRLACGVIG IAQ
Greater than 90% as determined by SDS-PAGE.
20655
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers