missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SOD1 / Superoxide Dismutase Protein
A DNA sequence encoding the Mus musculus (Mouse) SOD1 / Superoxide Dismutase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6715-ec-50ug
Additional Details : Weight : 0.01000kg
Spezifikation
NP_035564.1 | |
SOD1 / Superoxide Dismutase Protein | |
Research Use Only | |
sod1 | |
Recombinant Protein | |
E. coli | |
AMKAVCVLKG DGPVQGTIHF EQKASGEPVV LSGQITGLTE GQHGFHVHQY GDNTQGCTSA GPHFNPHSKK HGGPADEERH VGDLGNVTAG KDGVANVSIE DRVISLSGEH SIIGRTMVVH EKQDDLGKGG NEESTKTGNA GSRLACGVIG IAQ | |
Greater than 90% as determined by SDS-PAGE. |
20655 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |