missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SHH / Sonic hedgehog Protein

Product Code. 15983048
Click to view available options
Quantity:
100 μg
25 μg
250 μg
5 μg
Unit Size:
100µg
250µg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15983048

Brand: enQuireBio™ QP10194ec5ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mouse SHH / Sonic hedgehog, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_033196.1
Gene ID (Entrez) 20423
Name SHH / Sonic hedgehog Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol SHH
Protein Length Partial (see sequence information for more details).
Biological Activity The ED50 as determined by inducing alkaline phosphatase production of murine C3H10T1/2 cells is 0.5 - 1.0 ug/ml.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence IVIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKITRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGG
Buffer 0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.