missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse SEZ6/BSRP-C Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
50 μg
missing translation for 'unitSize'
50µg
Beschreibung
The Recombinant Mouse SEZ6/BSRP-C Protein is derived from NS0. The Recombinant Mouse SEZ6/BSRP-C Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
| Accession Number | Q7TSK2 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS |
| Gene ID (Entrez) | 20370 |
| Molecular Weight (g/mol) | 97kDa |
| Name | SEZ6/BSRP-C |
| Preparation Method | Reconstitute at 250μg/mL in sterile PBS. |
| Quality Control Testing | SDS-PAGE: 140-190kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse SEZ6/BSRP-C protein Gly19-Asn902,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur