missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SerpinE1 / PAI-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) SerpinE1 / PAI-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6672-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
SerpinE1 / PAI-1 Protein | |
Research Use Only | |
Serpine1 | |
Recombinant Protein | |
E. coli | |
TLPLRESHTA HQATDFGVKV FQQVVQASKD RNVVFSPYGV SSVLAMLQMT TAGKTRRQIQ DAMGFKVNEK GTAHALRQLS KELMGPWNKN EISTADAIFV QRDLELVQGF MPHFFKLFQT MVKQVDFSEV ERARFIINDW VERHTKGMIN DLLAKGAVDE LTRLVLVNAL YFSGQWKTPF LEASTHQRLF HKSDGSTVSV PMMAQSNKFN YTEFTTPDGL EYDVVELPYQ RDTLSMFIAA PFEKDVHLSA LTNILDAELI RQWKGNMTRL PRLLILPKFS LETEVDLRGP LEKLGMPDMF SATLADFTSL SDQEQLSVAQ ALQKVRIEVN ESGTVASSST AFVISARMAP TEMVIDRSFL FVVRHNPTET ILFMGQVMEP | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |