missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SERPINA9 Protein
A cDNA sequence encoding the SERPINA9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13460-50ug
Additional Details : Weight : 0.01000kg
Specifications
SERPINA9 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
NPYNQESSHL PSMKKNPASQ VSPSNTRFSF LLYQRLAQEN PGQNILFSPV SISTSLAMLS LGARSATKTQ ILRTLGFNFT WVSEPTIHMG FEYLVRSLNK CHQGRELRMG SVLFIRKELQ LQATFLDRVK KLYGAKVFSE DFSNAATAQA QINSYVEKET KGKVVDVIQD LDSQTAMVLV NHIFFKANWT QPFSTANTNK SFPFLLSKGT TVHVPMMHQT ESFAFGVDKE LGCSILQMDY RGDAVAFFVL PGKGKMRQLE KSLSARRLRK WSRSLQKRWI KVFIPKFSIS ASYNLETILP KMGIRDAFNS NADFSGITKT HFLQVSKAAH KAVLDVSEEG TEAAAATTTK LIVRSRDTPS SIIAFKEPFL ILLLDKNTES VLFLGKVENP RKMLEHHHHH H | |
Greater than 85.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
SERPINA9 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |