missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Serpin A1c/alpha-1-Antitrypsin Protein

Product Code. 18395527
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395527 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395527 Supplier R&D Systems™ Supplier No. 2979PI010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Serpin A1c/alpha-1-Antitrypsin Protein is derived from NS0. The Recombinant Mouse Serpin A1c/alpha-1-Antitrypsin Protein has been validated for the following applications: Inhibition Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in Sodium Acetate, NaCl and Glycerol.
Gene ID (Entrez) 20702
Molecular Weight (g/mol) 45 kDa
Name Serpin A1c/alpha 1-Antitrypsin
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse Serpin A1c/alpha 1-Antitrypsin protein Glu25-Lys413,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.