missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Semaphorin 3A Fc Chimera Protein

Product Code. p-4722141
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18379707

Brand: R&D Systems™ 5926S3025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Semaphorin 3A Fc Chimera Protein is derived from CHO. The Recombinant Mouse Semaphorin 3A Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Molecular Weight (g/mol) 110.1 kDa (monomer)
Name Semaphorin 3A Fc Chimera
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Semaphorin 3A protein, (N-terminus) Mouse Semaphorin 3A (Asn21-Lys747: Arg552Ala,Arg554Ala,Arg555Ala,Arg618Ala,Lys619Ala,Lys642Ala,Lys643Ala) Accession &Num; O08665 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.