missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Scrapie-responsive protein 1

A DNA sequence encoding the Mus musculus (Mouse) Scrapie-responsive protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7487-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008980

  • £560.00 / 100µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033162.1
Scrapie-responsive protein 1
Research Use Only
SCRG1
Recombinant Protein
Yeast
MPSSRLSCYR KLLKDRNCHN LPEGRADLKL IDANVQHHFW DGKGCEMICY CNFSELLCCP KDVFFGPKIS FVIPCNNH
Greater than 90% as determined by SDS-PAGE.
20284
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers