missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A9 Protein
A DNA sequence encoding the Mus musculus (Mouse) S100A9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6643-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001268781.1 | |
S100A9 Protein | |
Research Use Only | |
S100A9 | |
Recombinant Protein | |
E. coli | |
ANKAPSQMER SITTIIDTFH QYSRKEGHPD TLSKKEFRQM VEAQLATFMK KEKRNEALIN DIMEDLDTNQ DNQLSFEECM MLMAKLIFAC HEKLHENNPR GHGHSHGKGC GK | |
Greater than 90% as determined by SDS-PAGE. |
20202 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |