missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse S100A8/S100A9 Heterodimer Protein

Product Code. 18388947
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388947 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388947 Supplier R&D Systems™ Supplier No. 8916S8050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse S100A8/S100A9 Heterodimer Protein is derived from E. coli. The Recombinant Mouse S100A8/S100A9 Heterodimer Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Molecular Weight (g/mol) Observed M.W.: 7kDa and 12kDa, reducing conditions, Theoretical M.W.: 10kDa (S100A8), 13kDa (S100A9)
Quantity 50 μg
Source E. coli-derived mouse S100A8/S100A9 Heterodimer protein, (N-terminus) Mouse S100A8 (Pro2-Glu89) Accession &Num; P27005 Mouse S100A9 (Ala2-Lys113) Accession &Num; P31725 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name S100A8/S100A9 Heterodimer
Biological Activity Measured by its ability to induce IL-6 secretion by A375 human melanoma cells. Hibino, T. et al. (2013) Cancer Res. 73:172. The ED50 for this effect is typically 1.5-6μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System E. coli-derived Mouse S100A8 (Pro2-Glu89) Accession # P27005, N-terminal; Mouse S100A9 (Ala2-Lys113) Accession # P31725, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.