missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse S100A5 Protein

A cDNA sequence encoding the S100A5 was constructed and used to recombinantly synthesize the protein.

£98.39 - £2978.71

Specifications

Name S100A5 Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Mouse
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15963429
View Documents Promotion Details
enQuireBio™
QP13395-5UG
5 μg
£98.39
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15953429
View Documents Promotion Details
enQuireBio™
QP13395-20UG
20 μg
£151.37
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15943429
View Documents Promotion Details
enQuireBio™
QP13395-1MG
1 mg
£2978.71
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

S100A5 Protein
His
Mouse
Greater than 95.0% as determined by SDS-PAGE.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMETPLE KALTTMVTTF HKYSGREGSK LTLSRKELKE LIKTELSLAE KMKESSIDNL MKSLDKNSDQ EIDFKEYSVF LTTLCMAYND FFLEDNK
SDS
Product Certifications