missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse RSPO1 Protein
A cDNA sequence encoding the RSPO1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13365-100ug
Additional Details : Weight : 0.01000kg
Specifications
192199 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPSRGIKGK RQRRISAEGS QACAKGCELC SEVNGCLKCS PKLFILLERN DIRQVGVCLP SCPPGYFDAR NPDMNKCIKC KIEHCEACFS HNFCTKCQEG LYLHKGRCYP ACPEGSTAAN STMECGSPAQ CEMSEWSPWG PCSKKRKLCG FRKGSEERTR RVLHAPGGDH TTCSDTKETR KCTVRRTPCP EGQKRRKGGQ GRRENANRHP ARKNSKEPGS NSRRHKGQQQ PQPGTTGPLT SVGPTWAQHH HHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
RSPO1 Protein | |
Research Use Only | |
Rspo1 | |
Mouse | |
His | |
RSPO1 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |