missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse ROR2 mFc Chimera Protein, CF
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. Recombinant Mouse ROR2 mFc Chimera (Catalog # 11425-RO) binds Human ROR2 Antibody (Catalog # AF2064 ) with an ED50 of 0.600-7.20 ng/mL.
Specifications
Specifications
| Accession Number | NP_038874.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. |
| Gene ID (Entrez) | 4920 |
| Molecular Weight (g/mol) | M.W. (Observed): 80-88 kDa, under reducing conditions., M.W. (Theroretical): 68 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse ROR2 protein Mouse ROR2 (Glu34-Gly403) (N-terminus) Accession # NP_038874.3 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction