missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse RALA Protein
A cDNA sequence encoding the RALA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13257-5ug
Additional Details : Weight : 0.01000kg
Specifications
RALA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAANKP KGQNSLALHK VIMVGSGGVG KSALTLQFMY DEFVEDYEPT KADSYRKKVV LDGEEVQIDI LDTAGQEDYA AIRDNYFRSG EGFLCVFSIT EMESFAATAD FREQILRVKE DENVPFLLVG NKSDLEDKRQ VSVEEAKNRA DQWNVNYVET SAKTRANVDK VFFDLMREIR ARKMEDSKEK NGKKKRKSLA KRIRERC | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
RALA protein solution (0.5 mg/ml) 20mM Tris-Hcl buffer (pH 8.5) containing 30% glycerol, 0.1M NaCl and 0.1mM PMSF. |