missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse R-Spondin 4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£373.64
Specifications
Gene ID (Entrez) | 228770 |
---|---|
Molecular Weight (g/mol) | 23.9 kDa |
Name | R-Spondin 4 |
Quantity | 25μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse R-Spondin 4 protein Ala19-Gln228 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18156750
|
R&D Systems™
4106-RS-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£373.64
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18319687
|
R&D Systems™
4106-RS-025 |
£373.64
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse R-Spondin 4 Protein is derived from CHO. The Recombinant Mouse R-Spondin 4 Protein has been validated for the following applications: Bioactivity.Specifications
228770 | |
R-Spondin 4 | |
Chinese Hamster Ovary cell line,CHO-derived mouse R-Spondin 4 protein Ala19-Gln228 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
23.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |