missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Mouse R-Spondin 3 mFc Protein, CF

Product Code. 30040322
Click to view available options
Quantity:
10 μg
500 μg
Unit Size:
10µg
500µg
This item is not returnable. View return policy

Product Code. 30040322

Brand: R&DSystems™ 11540RS010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to activate TCF reporter activity in HEK293 human embryonic kidney cells in the presence of Wnt-3a. The ED50 for this effect is 0.300-3.60 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2TJ95.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 84870
Molecular Weight (g/mol) M.W.-observed: 54-65 kDa, under reducing conditions, M.W.-theroretical: 48 kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse R-Spondin 3 protein Mouse RSPO03 (Gln22, Met33-Gly209) (N-terminus) Accession # Q2TJ95.2 IEGRMD Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein R-Spondin 3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.