missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Ptprn2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Ptprn2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9357-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088181

  • £1600.00 / 500µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_035345.2
Ptprn2 Protein
Research Use Only
Ptprn2
Recombinant Protein
Yeast
RGRQLPGRLG CLFEDGLCGS LETCVNDGVF GRCQKVPVMD TYRYEVPPGA LLHLKVTLQK LSRTGFTWQD DYTQRVIAQE LANLPKAYLW HGEASGPARS LQQNADNEKW FSLEREVALA KTLRRYLPYL ELLSQTPTAN AHSRIDHETR PAKGEDSSPE NILTYVAHTS ALTYPPATRA KYPDNLLRPF SRLQPDELSP KVDGDIDKQK LIAALGAYTA QRLPGENDPE PRYLVHGSAR APRPFSATAL SQRWPPPPGD AKDSPSMDDD TLLQSLLKDL QQNSEVDRLG PLKEEKADSV AGAIQSDPAE GSQESHGRGA EGQPREQTDA PETMLQDHRL SEVDDPVYKE VNRLSFQLGD LLKDYGSPLL PEGPLLEKSS REEIKKSEQP EEVLSSEEET AGVEHVRSRT YSKDLFERKP NSEPQPRRLE DQFQNRAPEL WEDEESLKLA AQGPPSGGLQ LEVQPSEEQQ GYILTGNNPL SPEKGKQLMD QVAHILRVPS SFFADIKVLG PAVTFKVSAN IQNMTTADVI KAAADNKDQL EKATGLTILQ SGIRPKGKLK LLPHQEEQED STK
Greater than 90% as determined by SDS-PAGE.
19276
500 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers