missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Pentraxin 3/TSG-14 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
£345.56
Specifications
Gene ID (Entrez) | 19288 |
---|---|
Molecular Weight (g/mol) | 41 kDa (monomer) |
Name | Pentraxin 3/TSG-14 |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived mouse Pentraxin 3/TSG-14 protein Glu18-Ser381,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18126160
|
R&D Systems™
2166-TS-025 |
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein. |
£345.56
25µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18197910
|
R&D Systems™
2166-TS-025/CF |
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 . |
£345.56
25µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse Pentraxin 3/TSG-14 Protein is derived from NS0. The Recombinant Mouse Pentraxin 3/TSG-14 Protein has been validated for the following applications: Binding Activity.Specifications
19288 | |
Pentraxin 3/TSG-14 | |
Mouse myeloma cell line,NS0-derived mouse Pentraxin 3/TSG-14 protein Glu18-Ser381,with a C-terminal 6-His tag | |
Multimer consisting of as many as ten non-covalently and covalently linked subunits | |
Recombinant |
41 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |