missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PDIA3 Protein

A cDNA sequence encoding the PDIA3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10807-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965228

  • £155.00 / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

14827
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is >15 A650/cm/min/mg,obtained by measuring the increase of insulin precipitation in absorbance at 650nm resulting from the reduction of insulin.
Mouse
His
PDIA3 protein solution (1 mg/ml) containing 20 mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 10% glycerol .
PDIA3 Protein
Research Use Only
Pdia3
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSDVLEL TDENFESRVS DTGSAGLMLV EFFAPWCGHC KRLAPEYEAA ATRLKGIVPL AKVDCTANTN TCNKYGVSGY PTLKIFRDGE EAGAYDGPRT ADGIVSHLKK QAGPASVPLR TEEEFKKFIS DKDASVVGFF RDLFSDGHSE FLKAASNLRD NYRFAHTNIE SLVKEYDDNG EGITIFRPLH LANKFEDKTV AYTEKKMTSG KIKKFIQDSI FGLCPHMTED NKDLIQGKDL LTAYYDVDYE KNAKGSNYWR NRVMMVAKKF LDAGHKLNFA VASRKTFSHE LSDFGLESTT GEVPVVAIRT AKGEKFVMQE EFSRDGKALE QFLQEYFDGN LKRYLKSEPI PESNEGPVKV VVAENFDDIV NEEDKDVLIE FYAPWCGHCK NLEPKYKELG EKLSKDPNIV IAKMDATAND VPSPYEVKGF PTIYFSPANK KLTPKKYEGG RELNDFISYL QREATNPPII QEEKPKKKKK AQEDL
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers