missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse PD-L2/B7-DC Fc Chimera Protein

Product Code. 18749258
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18749258

Brand: R&D Systems™ 1022PL01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3-induced proliferation of stimulated mouse T cells. The ED50 for this effect is 1-6 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9WUL5
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 80380
Molecular Weight (g/mol) M.W. (Observed): 76 kDa, reducing conditions; M.W. (theoretical): 49 kDa (monomer)
Quantity 1 mg
Source Mouse myeloma cell line, NS0-derived mouse PD-L2/B7-DC protein Mouse PD-L2 (Leu20-Arg219) (N-terminus) Accession # Q9WUL5 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2,mgC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Gene Symbol Pdcd1lg2
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 400 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-L2/B7-DC/PDCD1LG2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.