missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Pcsk5 Protein

A DNA sequence encoding the Mus musculus (Mouse) Pcsk5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6474-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063560

  • £560.00 / 100µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001177412.1
Pcsk5 Protein
Research Use Only
Pcsk5
Recombinant Protein
Yeast
DYDLSHAQST YFNDPKWPSM WYMHCSDNTH PCQSDMNIEG AWKRGYTGKN IVVTILDDGI ERTHPDLMQN YDALASCDVN GNDLDPMPRY DASNENKHGT RCAGEVAATA NNSHCTVGIA FNAKIGGVRM LDGDVTDMVE AKSVSYNPQH VHIYSASWGP DDDGKTVDGP APLTRQAFEN GVRMGRRGLG SVFVWASGNG GRSKDHCSCD GYTNSIYTIS ISSTAESGKK PWYLEECSST LATTYSSGES YDKKIITTDL RQRCTDNHTG TSASAPMAAG IIALALEANP FLTWRDVQHV IVRTSRAGHL NANDWKTNAA GFKVSHLYGF GLMDAE
Greater than 90% as determined by SDS-PAGE.
18552
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers