missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse Park7/DJ-1 His Protein

Product Code. p-7111107 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100 μg
Unit Size:
0.50mg
100µg
This item is not returnable. View return policy

Product Code. 18282923

Brand: Novus Biologicals™ NBP2595230.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-189 of Mouse Park7/DJ-1 The Recombinant Mouse Park7/DJ-1 Protein is derived from E. coli. The Recombinant Mouse Park7/DJ-1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 20% glycerol,1mM DTT
Gene ID (Entrez) 11315
Molecular Weight (g/mol) 22.4 kDa
Name Recombinant Mouse Park7/DJ-1 Protein
Purification Method >90%, by SDS-PAGE
Quantity 0.5 mg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Gene Symbol PARK7
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.