missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Osteoprotegerin/TNFRSF11B Fc Chimera

Product Code. 18340408
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340408 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340408 Supplier R&D Systems™ Supplier No. 459MO100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit TRAIL-mediated cytotoxicity using L-929 mouse fibroblast cells treated with TRAIL. The ED50 for this effect is 3-15 ng/mL in the presence of 12 ng/mL of cross-linked Recombinant Human TRAIL/TNFSF10 (Catalog #s 375-TL and MAB050 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6PI12
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4982
Molecular Weight (g/mol) M.W. (Observed): 85-95 kDa, reducing conditions; M.W. (theoretical): 70.9 kDa (monomer)
Name Osteoprotegerin/TNFRSF11B Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived mouse Osteoprotegerin/TNFRSF11B protein Mouse OPG Glu22-Leu401 (Gln138Arg) (N-terminus) Accession # Q6PI12 IEGRMD Human IgG1 (Pro100-Lys330) (Thr276Ala) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tnfrsf11b
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Osteoprotegerin/TNFRSF11B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.