missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse NRG4 GST Protein

Product Code. 18325036 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325036 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325036 Supplier Novus Biologicals™ Supplier No. NBP318162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Should bind to mouse and human receptor tyrosine-protein kinase ErbB4 (according to literature)
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9WTX4
Concentration LYOPH
For Use With (Application) Bioactivity
Formulation Lyophilized from PBS
Gene ID (Entrez) 145957
Molecular Weight (g/mol) M.W. Theoretical: 32 kDa
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.01 EU/μg purified protein (LAL test; Lonza).
Gene Alias DKFZp779N0541, DKFZp779N1944, heregulin 4, HRG4pro-NRG4, neuregulin 4, pro-neuregulin-4, membrane-bound isoform, Pro-NRG4
Gene Symbol NRG4
Reconstitution Reconstitute with 100 ul sterile water (0.1mg/ml after reconstitution)
Research Category Cell Biology, Neuroscience, Signal Transduction
Purity or Quality Grade >95% pure by SDS-PAGE
Protein NRG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.