missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Mouse Noggin Protein
Klik for at se tilgængelige muligheder
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Pakningsstørrelse:
25µg
Beskrivelse
The Recombinant Mouse Noggin Protein is derived from NS0. The Recombinant Mouse Noggin Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 18121 |
| Molecular Weight (g/mol) | 26 kDa (monomer) |
| Name | Noggin |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Noggin protein, (N-terminus) Mouse Noggin (Leu20-Cys232) Accession &Num; P97466 IEGRGGGSGGGSGGGS 10-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A |
| Conjugate | Unconjugated |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion