missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Noggin (aa 28-232) Protein

Product Code. 18196120
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18196120 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 25µg
18358377 Lyophilized from a 0.2 μm filtered solution in PBS. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18196120 Supplier R&D Systems™ Supplier No. 6997NG025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.04-0.16 μg/mL in the presence of 50 ng/mL of Recombinant Human BMP-4 (Catalog # 314-GP ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P97466
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 9241
Molecular Weight (g/mol) M.W. (Observed): 25-35 kDa, reducing conditions; M.W. (theoretical): 23 kDa (monomer)
Name Noggin (aa 28-232)
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived mouse Noggin protein Gln28-Cys232
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A
Gene Symbol Nog
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Noggin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.