missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NNMT Protein
A cDNA sequence encoding the NNMT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12862-10ug
Additional Details : Weight : 0.01000kg
Specifications
18113 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMESGFT SKDTYLSHFN PRDYLEKYYS FGSRHCAENE ILRHLLKNLF KIFCLGAVKG ELLIDIGSGP TIYQLLSACE SFTEIIVSDY TDQNLWELQK WLKKEPGAFD WSPVVTYVCD LEGNRMKGPE KEEKLRRAIK QVLKCDVTQS QPLGGVSLPP ADCLLSTLCL DAACPDLPAY RTALRNLGSL LKPGGFLVMV DALKSSYYMI GEQKFSSLPL GWETVRDAVE EAGYTIEQFE VISQNYSSTT SNNEGLFSLV GRKPGRSE | |
Greater than 90.0% as determined by SDS-PAGE. |
NNMT Protein | |
Research Use Only | |
Nnmt | |
Mouse | |
His | |
NNMT protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0), 10% glycerol and 1mM DTT. |