missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Neutrophil Elastase/ELA2 Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Mouse Neutrophil Elastase/ELA2 Protein is derived from NS0. The Recombinant Mouse Neutrophil Elastase/ELA2 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 50701 |
| Molecular Weight (g/mol) | 27 kDa |
| Name | Neutrophil Elastase/ELA2 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Neutrophil Elastase/ELA2 protein Ser27-Arg260,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction