missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Mindin Protein

Product Code. 18706677
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18706677

Brand: R&D Systems™ 8500SP050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Mindin Protein is derived from NS0. The Recombinant Mouse Mindin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8BMS2
For Use With (Application) Bioactivity
Gene ID (Entrez) 10417
Molecular Weight (g/mol) Observed M.W.: 38-47kDa, reducing conditions, Theoretical M.W.: 35kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Mindin protein, (N-terminus) Mouse Mindin (Gln26-Val330) Accession &Num; Q8BMS2 HP GGGSGGGSGGGS HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Differentially expressed in cancerous and non-cancerous lung cells 1, DIL1, DIL-1, DIL1FLJ34460, DKFZp686G21139, FLJ16313, FLJ22401, MINDIN, M-SPONDIN, RG-1, SPON2, Spondin 2, spondin 2, extracellular matrix protein, spondin-2
Common Name Mindin/Spondin-2
Gene Symbol Spon2
Biological Activity Measured by its ability to bind fluorescein-conjugated E. coli Bioparticles. Sankala, M. et al. (2002) J. Biol. Chem. 277:33378. The ED50 for this effect is typically 0.3-1.2μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse Mindin (Gln26-Val330) Accession # Q8BMS2, N-terminus; HP GGGSGGGSGGGS HHHHHH, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.