missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Microtubule-associated protein 1

A DNA sequence encoding the Mus musculus (Mouse) Microtubule-associated protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6335-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052560

  • £560.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001297563.1
Microtubule-associated protein 1
Research Use Only
Map2
Recombinant Protein
Yeast
GMEGEKLPPV PFAQTFGTNL EDRKQSTEPS IVMPSIGLSA EPPAPKEPKD WFIEMPTESK KDEWGLAAPI SPGPLTPMRE KDVLEDIPRW EGKQFDSPMP SPFHGGSFTL PLDTMKNERV SEGPRPFAPV FFQSDDKVSL QDPSALATSK ESSKDEEPLK DKADKVADVS ISEVTTLLGN VHSPVVEGYV GENISGEVKV TTDQEKKETS APSVQEPTLT ETEPQTKLDE KSTVSIEEAV AKKEESFKLR DDKTGVIQTS TEQSFSKEDQ KGQEHTIDEL KQDSFPISLE QAVTDAAMTS KTLGKVTSEP EAVSERREIQ GLFEEKTADK NKLEGAGSAT IAEVEMP
Greater than 90% as determined by SDS-PAGE.
17756
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers