missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Meteorin-like/METRNL Protein
Description
The Recombinant Mouse Meteorin-like/METRNL Protein is derived from CHO. The Recombinant Mouse Meteorin-like/METRNL Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 210029 |
| Molecular Weight (g/mol) | 29.4 kDa |
| Name | Meteorin-like/METRNL |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Meteorin-like/METRNL protein Gln46-Glu311 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?