missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCP-2 / CCL8 Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_067418.1 |
---|---|
Gene ID (Entrez) | 20307 |
Name | MCP-2 / CCL8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966199
|
enQuireBio™
QP5471-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946199
|
enQuireBio™
QP5471-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976199
|
enQuireBio™
QP5471-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956199
|
enQuireBio™
QP5471-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_067418.1 | |
MCP-2 / CCL8 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
THP-1 chemotaxis significantly induced by less than 100 ng/ml of Mouse MCP-2 / CCL8. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
20307 | |
Research Use Only | |
Ccl8 | |
Recombinant Protein | |
E. coli | |
GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP | |
Greater than 95% as determined by SDS-PAGE. |