missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Mast Cell Protease-11/Prss34 Protein

Product Code. 18166290
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18166290

Brand: R&D Systems™ 2857SE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Mast Cell Protease-11/Prss34 Protein is derived from NS0. The Recombinant Mouse Mast Cell Protease-11/Prss34 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in MES and NaCl.
Gene ID (Entrez) 328780
Molecular Weight (g/mol) 34 kDa
Name Mast Cell Protease-11/Prss34
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse Mast Cell Protease-11/Prss34 protein Met20-Ser318, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Structural Form Pro form
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.