missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MAPT / Tau Protein

Product Code. 16077951
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16077951

Brand: enQuireBio™ QP9264ye500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) MAPT / Tau, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001033698.1
Gene ID (Entrez) 17762
Name MAPT / Tau Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Mapt
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Yeast
Protein Tag His
Sequence ADPRQEFDTM EDHAGDYTLL QDQEGDMDHG LKESPPQPPA DDGAEEPGSE TSDAKSTPTA EDVTAPLVDE RAPDKQAAAQ PHTEIPEGIT AEEAGIGDTP NQEDQAAGHV TQGRREGQAP DLGTSDWTRQ QVSSMSGAPL LPQGLREATC QPSGTRPEDI EKSHPASELL RRGPPQKEGW GQDRLGSEEE VDEDLTVDES SQDSPPSQAS LTPGRAAPQA GSGSVCGETA SVPGLPTEGS VPLPADFFSK VSAETQASQP EGPGTGPMEE GHEAAPEFTF HVEIKASTPK EQDLEGATVV GVPGEEQKAQ TQGPSVGKGT KEASLQEPPG KQPAAGLPGR PVSRVPQLKA RVASKDRTGN DEKKAKTSTP SCAKAPSHRP CLSPTRPTLG SSDPLIKPSS PAVSPEPATS PKHVSSVTPR NGSPGTKQMK LKGADGKTGA KIATPRGAAS PAQKGTSNAT RIPAKTTPSP KTPPGSGEPP KSGERSGYSS PGSPGTPGSR SRTPSLPTPP TREPKKVAVV RTPPKSPSAS KSRLQTAPVP MPDLKNVRSK IGSTENLKHQ PGGGKVQIIN KKLDLSNVQS KCGSKDNIKH VPGGGSVQIV YKPVDLSKVT SKCGSLGNIH HKPGGGQVEV KSEKLDFKDR VQSKIGSLDN ITHVPGGGNK KIETHKLTFR ENAKAKTDHG AEIVYKSPVV SGDTSPRHLS NVSSTGSIDM VDSPQLATLA DEVSASLAKQ GL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.