missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse MAG/Siglec-4a Fc Chimera Protein

Product Code. 18718247
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18718247

Brand: R&D Systems™ 8580MG100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse MAG/Siglec-4a Fc Chimera Protein is derived from NS0. The Recombinant Mouse MAG/Siglec-4a Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 4099
Molecular Weight (g/mol) Observed M.W.: 104-124kDa, reducing conditions, Theoretical M.W.: 82kDa (monomer)
Quantity 100 μg
Source Mouse myeloma cell line,NS0-derived mouse MAG/Siglec-4a protein, (N-terminus) Mouse MAG/Siglec-4a (Gly20-Pro516) Accession &Num; P20917 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias GMAsialic acid binding Ig-like lectin 4A, myelin associated glycoprotein, myelin-associated glycoprotein, sialic acid-binding immunoglobulin-like lectin 4A, Siglec4a, SIGLEC-4A, S-MAG
Common Name MAG/Siglec-4a
Gene Symbol Mag
Biological Activity Measured by its ability to inhibit neurite outgrowth of dissociated E13 chick embryonic dorsal root ganglia (DRG) neurons. When immobilized as a 3μL droplet containing 200 ng on a nitrocellulose coated microplate, recombinant mouse MAF Fc Chimera is able to significantly inhibit neurite outgrowth.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse MAG/Siglec-4a (Gly20-Pro516) Accession # P20917, N-terminus; IEGRMDP; Mouse IgG2a (Glu98-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.