missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse M-CSF Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1mg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
MCSF Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine M-NFS-60 cells is less than 2 ng/mL, corresponding to a specific activity of >5.0 x 10^5 IU/mg.
Specifications
Specifications
| Accession Number | Q3U4F9, Q3U4F9, Q3U4F9 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in 20 mM Tris, 500 mM NaCl, pH 7.4. |
| Gene ID (Entrez) | 1435 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 52 (homodimer) |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of M-CSF as determined by LAL method. |
| Gene Alias | colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930 |
| Gene Symbol | CSF1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction