missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse M-CSF / CSF-1 Protein
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
Specifications
Specifications
| Accession Number | NP_001107001.1 |
| Gene ID (Entrez) | 12977 |
| Name | M-CSF / CSF-1 Protein |
| Quantity | 500 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | CSF1 |
| Biological Activity | Shows bioactivity at a concentration of 10 ng/mL in the NFS-60 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction