missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Lysyl oxidase homolog 1 Protein

Product Code. p-7153051
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16082420

Brand: enQuireBio™ QP6316ye10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Lysyl oxidase homolog 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_034859.2
Gene ID (Entrez) 16949
Name Lysyl oxidase homolog 1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Loxl1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Yeast
Protein Tag His
Sequence RQAPSLPLPG RVGSDTVRGQ TRHPFGFGQV PDNWREVAVG DSTGMARART SVSQQRHGGS ASSSVSASAF ATTYRQPSPY PQQFPYPQAP FVNQYENYDP ASRTYEQGYV YYRGAGGGMG AGAAAVASAG VIYPFQPRAR YEDYGGGGGE EQPEYPAQGF YPAPERPYVP QPQPQPQPQP QPQPQPSDGL DRRYSHSLYN EGTPGFEQAY PDPSTDVSQA PAGAGGTYGG AGDPRLGWYP PYAANVPPEA YVPPRAVEPQ PPFRVLEPPY LPVRSSDAPS QGGERNGAQQ GRLSVGSVYR PNQNGRGLPD LVPDPNYVQA STYVQRAHLY SLRCAAEEKC LASTAYAPEA TDYDLRVLLR FPQRVKNQGT ADFLPNRPRH TWEWHSCHQH YHSMDEFSHY DLLDASTGKK VAEGHKASFC LEDSTCDFGN LKRYACTSHT QGLSPGCYDT YNADIDCQWI DITDVQPGNY ILKVHVNPKY IVLESDFTNN VVRCNIHYTG RYVSTTNCKI VQS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.