missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Lipocalin-2/NGAL Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Mouse Lipocalin-2/NGAL Protein is derived from NS0. The Recombinant Mouse Lipocalin-2/NGAL Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 16819 |
| Molecular Weight (g/mol) | 22 kDa |
| Name | Lipocalin-2/NGAL |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Lipocalin-2/NGAL protein Gln21-Asn200 with a C-terminal 10 His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Gene Alias | 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido