missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LILRB3 / LIR3 / ILT5 / CD85a Protein
A DNA sequence encoding the Mus musculus (Mouse) LILRB3 / LIR3 / ILT5 / CD85a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9252-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035225.2 | |
LILRB3 / LIR3 / ILT5 / CD85a Protein | |
Research Use Only | |
Lilrb3 | |
Recombinant Protein | |
Yeast | |
RRRHRGKFRK DVQKEKDLQL SSGAEEPITR KGELQKRPNP AAATQEESLY ASVEDMQTED GVELNSWTPP EEDPQGETYA QVKPSRLRKA GHVSPSVMSR EQLNTEYEQA EEGQGANNQA AESGESQDVT YAQLCSRTLR QGAAASPLSQ AGEAPEEPSV YATLAAARPE AVPKDMEQ | |
Greater than 90% as determined by SDS-PAGE. |
18733 | |
200 μg | |
Not determined. | |
Cytoplasmic Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |