missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse LIGHT/TNFSF14 Protein

Product Code. 18647356 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18647356

Brand: Novus Biologicals™ NBP2613325ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

LIGHT/TNFSF14 protein is fully biologically active when compared to standard. The ED50 as determined by a cytotoxicity assay using human HT-29 cells is less than 2 μg/mL, corresponding to a specific activity of > 500 IU/mg in the presence of murine anti-polyHistidine monoclonal antibody and rHuIFN-g.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH7.4.
Gene ID (Entrez) 8740
Molecular Weight (g/mol) MolecularWeight-theroretical: 18.4 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of LIGHT/TNFSF14 as determined by LAL method.
Gene Alias CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Gene Symbol TNFSF14
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in 100 mM HAC to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Apoptosis
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein LIGHT/TNFSF14
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.