missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse LIF Protein

Product Code. 18727237
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
This item is not returnable. View return policy

Product Code. 18727237

Brand: R&D Systems™ 8878LF025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse LIF Protein is derived from E. coli. The Recombinant Mouse LIF Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09056
For Use With (Application) Bioactivity
Gene ID (Entrez) 3976
Molecular Weight (g/mol) Observed M.W.: 20kDa, reducing conditions, Theoretical M.W.: 20kDa
Quantity 25 μg
Source E. coli-derived mouse LIF protein Pro25-Phe203
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Common Name LIF
Gene Symbol Lif
Biological Activity Measured by its ability to induce IL-6 secretion by M1 mouse myeloid leukemia cells. The ED50 for this effect is typically 0.1-0.6ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System E. coli-derived Pro25-Phe203
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.