missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse LIF, Animal-Free Protein
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
Description
Supports mouse ES cell propagation in chemically defined, feeder-free iPSC culture
Specifications
Specifications
| Accession Number | P09056.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from acetonitrile/TFA |
| Gene ID (Entrez) | 3976 |
| Molecular Weight (g/mol) | M.W. (Observed): Monomeric mouse LIF protein only, M.W. (Theroretical): 20 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived mouse LIF protein |
| Storage Requirements | Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction