missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse LIF, Animal-Free Protein
Description
Supports mouse ES cell propagation in chemically defined, feeder-free iPSC culture
Specifications
Specifications
| Accession Number | P09056.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from acetonitrile/TFA |
| Gene ID (Entrez) | 3976 |
| Molecular Weight (g/mol) | M.W. (Observed): Monomeric mouse LIF protein only, M.W. (Theroretical): 20 kDa |
| Quantity | 500 μg |
| Source | E. coli-derived mouse LIF protein |
| Storage Requirements | Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?