missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Leptin Protein
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Beschreibung
The Recombinant Mouse Leptin Protein is derived from E. coli. The Recombinant Mouse Leptin Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl. |
| Gene ID (Entrez) | 16846 |
| Molecular Weight (g/mol) | 16 kDa |
| Name | Leptin/OB |
| Quantity | 1 mg |
| Source | E. coli-derived mouse Leptin/OB protein Val22-Cys167,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Spot an opportunity for improvement?Share a Content Correction